Learn More
MSC 3M Safety Glasses, 2.00 Diopter, Clear Anti-Fog, Scratch-Resistant
Catalog No. 502835445
Encompass
This protective eyewear offers an athletic-inspired design, combining protection and affordability. It features a clear anti-fog lens with a +2.0 reading diopter to assist workers with small print or detailed tasks. An optional foam gasket helps limit eye exposure to nuisance dust and provides additional cushioning. A large wraparound lens ensures comprehensive eye protection and a wide viewing area, while contoured temples provide a comfortable and snug fit.
  • Integrated reading diopter assists with small print or detailed work
  • Optional foam gasket limits dust exposure and provides cushioning
  • Wraparound lens offers full eye protection and a wide viewing area
  • Contoured temples provide a comfortable and snug fit
  • Impact-resistant polycarbonate lens absorbs 99.9% UV rays
  • Anti-fog and anti-scratch lens coatings
  • Sporty frame design with cord control system
Catalog No. 50-283-5445 Supplier MSC Supplier No. 53808143
This protective eyewear offers an athletic-inspired design, combining protection and affordability. It features a clear anti-fog lens with a +2.0 reading diopter to assist workers with small print or detailed tasks. An optional foam gasket helps limit eye exposure to nuisance dust and provides additional cushioning. A large wraparound lens ensures comprehensive eye protection and a wide viewing area, while contoured temples provide a comfortable and snug fit.
  • Integrated reading diopter assists with small print or detailed work
  • Optional foam gasket limits dust exposure and provides cushioning
  • Wraparound lens offers full eye protection and a wide viewing area
  • Contoured temples provide a comfortable and snug fit
  • Impact-resistant polycarbonate lens absorbs 99.9% UV rays
  • Anti-fog and anti-scratch lens coatings
  • Sporty frame design with cord control system