Learn More
MSC 3M Magnifying Safety Glasses, 1.50 Diopter, Clear Lens, Anti-Fog, ANSI Z87.1
Catalog No. 502834923
Encompass
3M Magnifying Safety Glasses are designed to provide enhanced vision and protection in various work settings. Featuring a 1.50 diopter clear lens, these glasses come with anti-fog and scratch-resistant coatings for optimal clarity and durability. The half-framed, wrap-around design, along with blue frames, ensures comfort, UV protection, and a secure fit.
  • 1.50 diopter lens for magnification
  • Clear lenses for unobstructed vision
  • Anti-fog coating improves visibility in humid conditions
  • Scratch-resistant lenses for enhanced durability
  • Blue frames and half-framed design
  • UV protection for eye safety
  • Wrap around frame design for secure fit
  • Non-adjustable temples for consistent wear
  • Compliant with ANSI Z87.1 and CSA Z94.3 safety standards
Catalog No. 50-283-4923 Supplier MSC Supplier No. 53808168
3M Magnifying Safety Glasses are designed to provide enhanced vision and protection in various work settings. Featuring a 1.50 diopter clear lens, these glasses come with anti-fog and scratch-resistant coatings for optimal clarity and durability. The half-framed, wrap-around design, along with blue frames, ensures comfort, UV protection, and a secure fit.
  • 1.50 diopter lens for magnification
  • Clear lenses for unobstructed vision
  • Anti-fog coating improves visibility in humid conditions
  • Scratch-resistant lenses for enhanced durability
  • Blue frames and half-framed design
  • UV protection for eye safety
  • Wrap around frame design for secure fit
  • Non-adjustable temples for consistent wear
  • Compliant with ANSI Z87.1 and CSA Z94.3 safety standards