Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Human B Cell Enrichment Kit (2nd Generation)

Catalog No. 8804686774
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-046-86774 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-046-86774 Supplier Invitrogen™ Supplier No. 8804686774
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The MagniSort™ Human B cell Enrichment Kit (2nd Generation) is designed for the magnetic separation of B cells by negative selection with improved purity.

The MagniSort™ Human B cell Enrichment Kit (2nd Generation) is designed for the magnetic separation of B cells by negative selection with improved purity. It has been optimized for the isolation of human B cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave B cells untouched and free in solution.

The MagniSort™ Human B cell Enrichment Antibody Cocktail (2nd Generation) contains the following antibodies:

  • Anti-Human CD2 Biotin
  • Anti-Human CD3 Biotin
  • Anti-Human CD11b Biotin
  • Anti-Human CD14 Biotin
  • Anti-Human CD16 Biotin
  • Anti-Human CD36 Biotin
  • Anti-Human CD56 Biotin
  • Anti-Human CD235a Biotin

Reported Application: Magnetic Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Cell Type B Cells
For Use With (Application) Magnetic Cell Separation
Product Type Cell Enrichment Kit
Format Kit
No. of Tests 200 Tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.