Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Human CD19 Positive Selection Kit

Catalog No. 8802683374
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-026-83374 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-026-83374 Supplier Invitrogen™ Supplier No. 8802683374
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MagniSort™ Human CD19 Positive Selection Kit is designed for the magnetic separation of CD19+ cells by positive selection.

The MagniSort™ Human CD19 Positive Selection Kit is designed for the magnetic separation of CD19+ cells by positive selection. It has been optimized for the isolation of CD19+ cells from normal human peripheral blood mononuclear cells, utilizing a biotinylated Anti-Human CD19 antibody and streptavidin-coated magnetic beads. CD19+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD19+ cells by decanting.

After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD20, clone 2H7.

Reported Application: Magnetic Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD19+
For Use With (Application) Magnetic Cell Separation
Product Type Positive Selection Kit
Format Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.