Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Human CD4 Naïve T cell Enrichment Kit

Catalog No. 8804681474
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-046-81474 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 88-046-81474 Supplier Invitrogen™ Supplier No. 8804681474
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide

Includes

  • MagniSort Human CD4 Naïve T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads A

Designed for the magnetic separation of CD4 naïve T cells by negative selection. It has been optimized for the isolation of human CD4 naïve T cells from normal human peripheral blood mononuclear cells utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 naïve T cells untouched and free in solution.

The MagniSort™ Human CD4 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD8 Biotin
Anti-Human CD11b Biotin
Anti-Human CD14 Biotin
Anti-Human CD16 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD36 Biotin
Anti-Human CD45RO Biotin
Anti-Human CD56 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
Anti-Human γδ TCR Biotin
TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (Naïve, CD4)
Product Type Cell Enrichment Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human
I need to further purify my naive human T cells that have been isolated using the MagniSort Human CD4 Naive T cell Enrichment Kit (Cat. No. 8804-6814-74). Which purification kit can I use?

The CD4 naive T cells that are isolated using the MagniSort Human CD4 Naive T cell Enrichment Kit are untouched and free in solution. Hence, you can use any other purification kit afterwards.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.