Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Human CD4 T cell 2-Step Enrichment Kit

Catalog No. 8802683174
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-026-83174 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-026-83174 Supplier Invitrogen™ Supplier No. 8802683174
Add to Cart
Edge
Add to Cart

The MagniSort™ Human CD4 2-Step Enrichment Kit is designed for the magnetic separation of CD4+ T cells by positive selection after first depleting monocytes.

The MagniSort™ Human CD4 2-Step Enrichment Kit is designed for the magnetic separation of CD4+ T cells by positive selection after first depleting monocytes. It has been optimized for the isolation of CD4+ T cells from human peripheral blood monocuclear cells, utilizing biotinylated Anti-Human CD14 and Anti-Human CD4 antibodies, as well as streptavidin-coated magnetic beads. In the first step, monocytes are bound by Anti-Human CD14 antibody and then magnetic beads. When placed in a magnetic field, the monocytes are held in the magnet while the remaining cells are decanted and used for the second step. In the second step, the monocyte-depleted cells are bound by Anti-Human CD4 antibody and then magnetic beads. Now, when placed in the magnetic field, the CD4+ T cells are held in the magnet while undesired cells can be decanted.

After enrichment, the purity of selected cells can be verified by staining with Anti-Human CD4, clones OKT4 or SK3.

Reported Application: Magnetic Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD4+
For Use With (Application) Magnetic Cell Separation
Product Type Cell Enrichment Kit
Format Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.