Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Human CD56 Positive Selection Kit

Catalog No. 8802683574
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-026-83574 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 88-026-83574 Supplier Invitrogen™ Supplier No. 8802683574
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide

Includes

  • MagniSort Anti-Human CD56 Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A

Designed for the magnetic separation of CD56+ cells by positive selection. It has been optimized for the isolation of CD56+ cells from human normal peripheral blood mononuclear cells utilizing a biotinylated Anti-human CD56 antibody and streptavidin-coated magnetic beads.

  • CD56+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD56+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD56 (clone CMSSB)
TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD56+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.