Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Human pan-Monocyte Enrichment Kit

Catalog No. 8804683774
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-046-83774 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 88-046-83774 Supplier Invitrogen™ Supplier No. 8804683774
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide

Includes

  • MagniSort Human pan-Monocyte Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic separation of conventional CD14+ and unconventional CD16+CD14- monocytes by negative selection.

  • Undesired cells, including CD16+ NK cells, are bound by antibody and then magnetic beads
  • When placed in a magnetic field, monocytes are untouched and free in solution


The MagniSort™ Human pan-Monocyte Enrichment Antibody Cocktail contains the following antibodies:
Anti-Human CD2 Biotin
Anti-Human CD3 Biotin
Anti-Human CD7 Biotin
Anti-Human CD19 Biotin
Anti-Human CD20 Biotin
Anti-Human CD56 Biotin
Anti-Human CD57 Biotin
Anti-Human CD123 Biotin
Anti-Human CD235a Biotin
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Monocytes (pan)
Product Type Human Pan-Monocyte Enrichment Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.