Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Mouse CD11c Positive Selection Kit

Catalog No. 8802686174
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-026-86174 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 88-026-86174 Supplier Invitrogen™ Supplier No. 8802686174
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide

Includes

  • MagniSort™ Mouse CD11c Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads B

Designed for the magnetic separation of CD11c+ cells by positive selection. It has been optimized for the isolation of CD11c+ cells from mouse spleens utilizing a biotinylated Anti-Mouse CD11c antibody and streptavidin-coated magnetic beads.

  • CD11c+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD11c+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD11c, clone N418 and Anti-Mouse CD11b, clone M1/70
  • To reduce non-specific binding, the MagniSort™ Anti-Mouse CD11c Biotin contains Fc Receptor block
TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD11c+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.