Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit

Catalog No. 8802681774
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-026-81774 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-026-81774 Supplier Invitrogen™ Supplier No. 8802681774
Add to Cart
Add to Cart

The MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit is designed for the magnetic separation of CD335+ cells by positive selection.

The MagniSort™ Mouse CD335 (NKp46) Positive Selection Kit is designed for the magnetic separation of CD335+ cells by positive selection. It has been optimized for the isolation of CD335+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD335+ antibody and streptavidin-coated magnetic beads. CD335+ cells are bound by antibody and then magnetic beads. When placed in a magnetic field, the undesired cells can be separated from CD335+ cells by decanting.

After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD49b (Integrin alpha 2), clone DX5, or Anti-Mouse NK1.1, clone PK136, in C57BL, FVB/N and NZB mice.

Reported Application: Magnetic Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD335+
For Use With (Application) Magnetic Cell Separation
Product Type Positive Selection Kit
Format Kit
No. of Tests 200 Tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.