Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Mouse CD45.1 Positive Selection Kit

Catalog No. 501125077
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-5077 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 50-112-5077 Supplier Invitrogen™ Supplier No. 8802684874
  • $994.00
    $745.65 / Each of 1

    Save $248.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide

Includes

  • MagniSort™ Anti-Mouse CD45.1 Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads A

Designed for the magnetic separation of CD45.1+ cells by positive selection. It has been optimized for the isolation of CD45.1+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD45.1 antibody and streptavidin-coated magnetic beads.

  • CD45.1+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD45.1+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD45.1, clone A20
  • When the starting frequency of CD45.1 cells is greater than 75% of the total cells, the amount of MagniSort™ Magnetic Positive Selection Beads to be used can be increased to 40 μL/test to maximize recovery
TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD45.1+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.