Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Mouse CD45 Depletion Kit

Catalog No. 501125092
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-5092 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 50-112-5092 Supplier Invitrogen™ Supplier No. 8804686474
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide

Includes

  • MagniSort Anti-Mouse CD45 Biotin A : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic depletion of mouse CD45+ cells from mouse spleens, bone marrow or digested lung tissue. It uses a biotinylated Anti-Mouse CD45 antibody and streptavidin-coated magnetic beads.

  • CD45+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the CD45- cells can be separated from CD45+ cells by decanting
  • After depletion, the purity of enriched CD45- cells can be verified by staining with Anti-Mouse CD45, clone 30-F11
TRUSTED_SUSTAINABILITY

Specifications

Cell Type CD45+
Product Type Cell Depletion Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.