Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit

Catalog No. 8804682574
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-046-82574 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 88-046-82574 Supplier Invitrogen™ Supplier No. 8804682574
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD8 naïve T cells by negative selection.

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD8 naïve T cells by negative selection. It has been optimized for the isolation of mouse CD8 naïve T cells from spleens or lymphnodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 naïve T cells untouched and free in solution.

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:

  • Anti-Mouse CD4 Biotin
  • Anti-Mouse CD11b Biotin
  • Anti-Mouse CD19 Biotin
  • Anti-Mouse CD24 Biotin
  • Anti-Mouse CD44 Biotin
  • Anti-Mouse CD45R (B220) Biotin
  • Anti-Mouse CD49b (Integrin alpha 2) Biotin
  • Anti-Mouse Ly-6G (Gr-1) Biotin
  • Anti-Mouse γδ TCR Biotin
  • Anti-Mouse TER-119 Biotin

Reported Application: Magnetic Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (Naïve, CD8+)
For Use With (Application) Magnetic Cell Separation
Product Type Cell Enrichment Kit
Format Kit
No. of Tests 200 Tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.