Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Magnoflorine chloride | 6681-18-1 | 99.3% | 377.86 | C20H24ClNO4 | 10 MG
SDP

Catalog No. 5000296262
Encompass_Preferred

(+)-Magnoflorine chloride is an aporphine alkaloid chloride salt reported to reduce Candida albicans biofilm formation and to exhibit anti-fungal, anti-diabetic, and antioxidant activity. It is supplied as a high-purity research chemical for in vitro studies and analytical use.

  • High purity (≈99.3%).
  • Molecular formula C20H24ClNO4.
  • Molecular weight 377.86.
  • CAS number 6681-18-1.
  • Provided in small milligram quantities for research use.
  • Chloride salt form for improved stability and handling.

Catalog No. 50-002-96262 Supplier Medchemexpress LLC Supplier No. HYN053510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Magnoflorine chloride is an aporphine alkaloid chloride salt reported to reduce Candida albicans biofilm formation and to exhibit anti-fungal, anti-diabetic, and antioxidant activity. It is supplied as a high-purity research chemical for in vitro studies and analytical use.

  • High purity (≈99.3%).
  • Molecular formula C20H24ClNO4.
  • Molecular weight 377.86.
  • CAS number 6681-18-1.
  • Provided in small milligram quantities for research use.
  • Chloride salt form for improved stability and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.