Learn More
Apexbio Technology LLC MAGNOFLORINE-IODIDE-10MG
SDP
Catalog No. 5000570753
Encompass_Preferred
( )-Magnoflorine iodide (CAS No 4277-43-4) is a quaternary alkaloid primarily investigated in the field of pharmacology for its potential therapeutic effects Originating from traditional medicinal plants like Magnolia species it acts by influencing various biological pathways including modulation of calcium channels and antioxidant activity In vitro studies typically demonstrate efficacy within the nanomolar to low micromolar range indicating its potency across different assays Commonly used in cell culture models and animal studies ( )-Magnoflorine iodide is explored for its neuroprotective and anti-inflammatory properties making it a candidate in drug screening for conditions such as neurodegenerative diseases and inflammatory disorders Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70753 Supplier Apexbio Technology LLC Supplier No. N285110
( )-Magnoflorine iodide (CAS No 4277-43-4) is a quaternary alkaloid primarily investigated in the field of pharmacology for its potential therapeutic effects Originating from traditional medicinal plants like Magnolia species it acts by influencing various biological pathways including modulation of calcium channels and antioxidant activity In vitro studies typically demonstrate efficacy within the nanomolar to low micromolar range indicating its potency across different assays Commonly used in cell culture models and animal studies ( )-Magnoflorine iodide is explored for its neuroprotective and anti-inflammatory properties making it a candidate in drug screening for conditions such as neurodegenerative diseases and inflammatory disorders Experimental concentrations typically depend on the specific research design