Learn More
Cayman Chemical MagnolIae flos asrInIn 10mg
SDP
Catalog No. 502941293
Encompass_Preferred
A lignan; an inhibitor of Δ5-desaturase (Ki = 0.28 mM); selective for Δ5-desaturase over Δ6- and Δ9-desaturase; cytotoxic to A2780 and SKOV3 ovarian cancer cells (IC50s = 38.45 and 60.87 µM, respectively) but not immortalized ovarian surface epithelial cells (IC50 = >200 µM)
Catalog No. 50-294-1293 Supplier Cayman Chemical Supplier No. 2745610 MG
A lignan; an inhibitor of Δ5-desaturase (Ki = 0.28 mM); selective for Δ5-desaturase over Δ6- and Δ9-desaturase; cytotoxic to A2780 and SKOV3 ovarian cancer cells (IC50s = 38.45 and 60.87 µM, respectively) but not immortalized ovarian surface epithelial cells (IC50 = >200 µM)