Learn More
Medchemexpress LLC HY-N0163 10mg Medchemexpress, Magnolol CAS:528-43-8 Purity:>98%
SDP
Catalog No. 501933437
Encompass_Preferred
Medchemexpress, HY-N0163 10mg Magnolol CAS:528-43-8 Magnolol, a natural lignan isolated from the stem bark of Magnolia officinalis, is a dual agonist of both RXRα and PPARγ , with EC 50 values of 10.4 µM and 17.7 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3437 Supplier Medchemexpress LLC Supplier No. HYN016310MG
Medchemexpress, HY-N0163 10mg Magnolol CAS:528-43-8 Magnolol, a natural lignan isolated from the stem bark of Magnolia officinalis, is a dual agonist of both RXRα and PPARγ , with EC 50 values of 10.4 µM and 17.7 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.