Learn More
Selleck Chemical LLC Magnolol 10mg 528-43-8 NSC 293099
SDP
Supplier:  Selleck Chemical LLC S232110MG
Encompass_Preferred
Magnolol (NSC 293099) is a bioactive lignin found in the bark of the Houpu magnolia (Magnolia officinalis) which shows antifungal properties. It can block TNF-?-induced NF-KB activation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-567-2