Learn More
Enzo Life Sciences Magnolol (25mg). CAS: 528-43-8
SDP
Supplier:  Enzo Life Sciences ALX350352M025
Encompass_Preferred
Potent and highly tolerable antitumor and antiangiogenic compound. Anxiolytic, anti-thrombotic and antibacterial. Shows central depressant effects in vivo. Inhibitor of nitric oxide (NO) and TNF-α production in LPS-activated macrophages by the suppression of inducible nitric oxide synthase (iNOS; NOS II) expression (IC50=16.8µM). Isomeric with honokiol (ALX-350-350). Alternative name: 5,5'-Diallyl-2,2'-biphenyldiol. Purity: ≥98% (HPLC). Appearance: White to light yellow powder. Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8856