Learn More
Medchemexpress LLC Magnolol | 528-43-8 | 266.33 | 1 ML
SDP
Catalog No. 5000427411
Encompass_Preferred
Magnolol is a natural lignan isolated from the stem bark of *Magnolia officinalis*. It acts as a dual agonist of both RXRα and PPARγ, with EC50 values of 10.4 μM and 17.7 μM, respectively.
  • Natural product
  • Isolated from the stem bark of *Magnolia officinalis*
  • Dual agonist of RXRα and PPARγ
Catalog No. 50-004-27411 Supplier Medchemexpress LLC Supplier No. HYN016310MM/1ML
Magnolol is a natural lignan isolated from the stem bark of *Magnolia officinalis*. It acts as a dual agonist of both RXRα and PPARγ, with EC50 values of 10.4 μM and 17.7 μM, respectively.
  • Natural product
  • Isolated from the stem bark of *Magnolia officinalis*
  • Dual agonist of RXRα and PPARγ