Learn More
ALPAQUA ENGINEERING LLC MAGNUM EX UNIVERSAL PLATE
Catalog No. NC0482875
Encompass
The Magnum EX Universal Magnet plate contains 96 of our strongest ring-shaped Neodymium Iron-Boron (NeFeB) magnets, unique corner brackets and an elevated platform providing for universal 96-well plate and robotic platform compatibility.
Catalog No. NC0482875 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000380
The Magnum EX Universal Magnet plate contains 96 of our strongest ring-shaped Neodymium Iron-Boron (NeFeB) magnets, unique corner brackets and an elevated platform providing for universal 96-well plate and robotic platform compatibility.