Learn More
ALPAQUA ENGINEERING LLC MAGNUM FLX ENHNCED UNI MAG PLT
Catalog No. NC0802088
Encompass
The Magnum FLX Enhanced Universal Magnet Plate contains 96 of our strongest magnets in a solid-core shape for increased speed. This magnet plate is universally compatible with all commonly used 96-well microplates and robots.
Catalog No. NC0802088 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000400
The Magnum FLX Enhanced Universal Magnet Plate contains 96 of our strongest magnets in a solid-core shape for increased speed. This magnet plate is universally compatible with all commonly used 96-well microplates and robots.