Learn More
Description
- Provides three times greater brightness compared to traditional luminol reagent
- Produces Longer-lasting luminescence.
- Visible to the naked eye in Semi-dark conditions.
- Ideal for miscellaneous items, surfaces, or clothing that have been washed
- Mix in a spray bottle containing 125 mL of distilled water
- Makes invisible bloodstains produce an intense blue chemiluminescence
- Does not adversely affect DNA
- Operates within a wavelength range of 420 to 440 nm.
- Fully soluble
- Odorless
Specifications
Specifications
| Product Type | Bloodstain Reagent |
| For Use With (Application) | Latent Bloodstain Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
