Learn More
Aviva Systems Biology Corp MAGOH Antibody - N-terminal region (ARP60300_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60300_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGOH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MAGOH. Alternate names: MAGOH1, MAGOHAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-000