Learn More
Aviva Systems Biology Corp MAGOHB Antibody - C-terminal region (ARP62679_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62679_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAGOHB. It was validated on Western Blot. . Alternate names: FLJ10292, MGN2, mago, magohApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-736