Learn More
ABclonal Technology MAGP-2/MFAP5 Rabbit mAb
SDP
Catalog No. 5000005076
Encompass_Preferred
This gene encodes a 25-kD microfibril-associated glycoprotein which is a component of microfibrils of the extracellular matrix The encoded protein promotes attachment of cells to microfibrils via alpha-V-beta-3 integrin Deficiency of this gene in mice results in neutropenia Alternate splicing results in multiple transcript variants encoding different isoforms
Catalog No. 50-000-05076 Supplier ABclonal Technology Supplier No. A2352920UL
This gene encodes a 25-kD microfibril-associated glycoprotein which is a component of microfibrils of the extracellular matrix The encoded protein promotes attachment of cells to microfibrils via alpha-V-beta-3 integrin Deficiency of this gene in mice results in neutropenia Alternate splicing results in multiple transcript variants encoding different isoforms