Learn More
Invitrogen™ MAGPIX™ Drive Fluid Plus
Catalog No. 4050030
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 x 700 mL
Catalog No. Quantity
40-500-30 4 x 700 mL
1 options
Catalog No. 40-500-30 Supplier Invitrogen™ Supplier No. 4050030
Add to Cart
Add to Cart
MAGPIX Drive Fluid Plus is the delivery medium for transporting the sample being analyzed on the Luminex MAGPIX system to the optic components.
MAGPIX Drive Fluid Plus is the delivery medium for transporting the sample being analyzed on the Luminex MAGPIX system to the optic components. Each unit in the 4-pack of drive fluid contains 700 mL.
Please refer to the Luminex instrument user manual for proper installation and use of MAGPIX Drive Fluid Plus with the MAGPIX System.
This reformulated sheath product replaces the original MAGPIX Drive Fluid (Cat. No. MPXDF4PK). The new sheath formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
For Research Use Only. Not for use in diagnostic procedures. Not intended for human or animal therapeutic or diagnostic use
TRUSTED_SUSTAINABILITY
Specifications
Product Line MAGPIX
Quantity 4 x 700 mL
Will there be any differences in performance of my instrument when switching over to MAGPIX Drive Fluid Plus from the original MAGPIX Drive Fluid?
In-house testing has been done to confirm that assays deliver expected results when using MAGPIX Drive Fluid Plus. White Paper is available upon request from Technical Support (techsupport@thermofisher.com).
Are there special instructions or considerations when switching over to MAGPIX Drive Fluid Plus from the original MAGPIX Drive Fluid?
Yes, there are instructions available for flushing the instrument and replacing the drive fluid. Please contact Technical Support at techsupport@thermofisher.com for these instructions.
What is the advantage of MAGPIX Drive Fluid Plus over the original MAGPIX Drive Fluid?
MAGPIX Drive Fluid Plus (Cat. No. 4050030) replaces the original MAGPIX Drive Fluid (Cat. No. MPXDF4PK). The new sheath formulation is updated from a single preservative to a mixture of three preservatives, specifically, sodium azide, sodium benzoate and diazolidinyl urea. This allows for greater transit stability and reduces the likelihood of future acquired microbial resistance.