Learn More
ALPAQUA ENGINEERING LLC MAGPLATE 24
Catalog No. NC0482874
Encompass
The Alpaqua 24-well magnet plate contains integrated Spring Cushion Technology and a unique ring structure. It is designed for use with 24-well large volume (10ml) Whatman UniPlates.
Catalog No. NC0482874 Supplier ALPAQUA ENGINEERING LLC Supplier No. A000270
The Alpaqua 24-well magnet plate contains integrated Spring Cushion Technology and a unique ring structure. It is designed for use with 24-well large volume (10ml) Whatman UniPlates.