Learn More
Sigma Aldrich Fine Chemicals Biosciences MagRackTM 6 Cytiva 28 9489
Catalog No. 502904759
Encompass
MagRack(TM) 6 is designed for small-scale sample preparation using magnetic beads and enables handling of up to six samples in 1.5 ml microcentrifuge tubes.MagRack(TM) 6 is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads. Each rack consists of an anodized aluminum housing (blue) with a detachable plastic bar (white) containing a neodymium magnet and this design allows for optimal performance during protein purifications or enrichments. MagRack(TM) 6 is for sample volumes of up to 1.5 ml while MagRack(TM) Maxi can be used for sample volumes of up to 50 ml. The racks make an excellent complement to the following products (containing magnetic beads) from Cytiva Protein A Mag Sepharose(R) Protein A Mag Sepharose(R) Xtra ProteinG Mag Sepharose(R) Protein G Mag Sepharose(R) Xtra His Mag [TMSepharose(R)] Ni NHS Mag Sepharose(R) TiO2 Mag Sepharose(R) and Streptavidin Mag Sepharose(R).One of the main advantages of the magnetic beads separat
Catalog No. 50-290-4759 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28948964
MagRack(TM) 6 is designed for small-scale sample preparation using magnetic beads and enables handling of up to six samples in 1.5 ml microcentrifuge tubes.MagRack(TM) 6 is a magnetic rack for small-scale protein purification and sample enrichment with magnetic beads. Each rack consists of an anodized aluminum housing (blue) with a detachable plastic bar (white) containing a neodymium magnet and this design allows for optimal performance during protein purifications or enrichments. MagRack(TM) 6 is for sample volumes of up to 1.5 ml while MagRack(TM) Maxi can be used for sample volumes of up to 50 ml. The racks make an excellent complement to the following products (containing magnetic beads) from Cytiva Protein A Mag Sepharose(R) Protein A Mag Sepharose(R) Xtra ProteinG Mag Sepharose(R) Protein G Mag Sepharose(R) Xtra His Mag [TMSepharose(R)] Ni NHS Mag Sepharose(R) TiO2 Mag Sepharose(R) and Streptavidin Mag Sepharose(R).One of the main advantages of the magnetic beads separat