Learn More
ESI SOURCE SOLUTIONS MAGRESYN SAX 10ML
Catalog No. NC2544489
Encompass
For rapid protein fractionation of proteins by ion exchange & concentration and clean-up of proteins from dilute solutions pre-MS analysis
Catalog No. NC2544489 Supplier ESI SOURCE SOLUTIONS Supplier No. MRSAX010
For rapid protein fractionation of proteins by ion exchange & concentration and clean-up of proteins from dilute solutions pre-MS analysis