Learn More
ESI SOURCE SOLUTIONS MagReSyn SAX magnetic microparticles, 2 ml, strong anion exchange (SAX), 20 mg/ml concentration, 20% ethanol medium, binding capacity ≥ 20 mg/ml (BSA), particle size ~5-10 μm
Catalog No. NC2333622
Encompass
MagReSyn SAX magnetic microparticles are high-performance strong anion exchange polymeric supports designed for efficient capture and purification of negatively charged biomolecules. These microparticles excel in fractionating complex biological mixtures, making them ideal for preparations prior to advanced analytical techniques such as mass spectrometry and HPLC.
  • High performance strong anion exchange microparticles
  • Efficient for biomolecule capture and purification
  • Exceptional binding capacity of ≥ 20 mg/ml (BSA)
  • Rapid magnetic separation in under 10 seconds
  • Suitable for serum, plasma, urine, and cell lysates
  • Storage temperature range of 4-8°C
  • Multi-stability in pH range of 3.5-10
  • Volume of 2 ml with a concentration of 20 mg/ml
  • Particle size approximately 5-10 μm
Catalog No. NC2333622 Supplier ESI SOURCE SOLUTIONS Supplier No. MRSAX002
MagReSyn SAX magnetic microparticles are high-performance strong anion exchange polymeric supports designed for efficient capture and purification of negatively charged biomolecules. These microparticles excel in fractionating complex biological mixtures, making them ideal for preparations prior to advanced analytical techniques such as mass spectrometry and HPLC.
  • High performance strong anion exchange microparticles
  • Efficient for biomolecule capture and purification
  • Exceptional binding capacity of ≥ 20 mg/ml (BSA)
  • Rapid magnetic separation in under 10 seconds
  • Suitable for serum, plasma, urine, and cell lysates
  • Storage temperature range of 4-8°C
  • Multi-stability in pH range of 3.5-10
  • Volume of 2 ml with a concentration of 20 mg/ml
  • Particle size approximately 5-10 μm