Learn More
ESI SOURCE SOLUTIONS MAGRESYN STREPTAVIDIN -10 ML
Catalog No. NC1873311
Encompass
MagReSyn® Streptavidin - 10mlMagReSyn® Streptavidin microparticles contain covalently coupled streptavidin. The porous nature of the polymer technology allows for exceptional streptavidin capacity, translating into increased capacity for the immobilization/capture of biotinylated biomolecules. The high streptavidin load allows the use of as little as 10 µl beads, improving experimental miniaturization for high-throughput automated analysis. Molecular applications of MagReSyn® Streptavidin microparticles include, protein immobilization and purification; nucleic acid capture assays and preparation of single-stranded DNA templates for e.g. SELEX and pyrosequencing. The immobilization of biotinylated antibodies may subsequently be used in e.g. immunoassay.
Catalog No. NC1873311 Supplier ESI SOURCE SOLUTIONS Supplier No. MRSTV010
MagReSyn® Streptavidin - 10mlMagReSyn® Streptavidin microparticles contain covalently coupled streptavidin. The porous nature of the polymer technology allows for exceptional streptavidin capacity, translating into increased capacity for the immobilization/capture of biotinylated biomolecules. The high streptavidin load allows the use of as little as 10 µl beads, improving experimental miniaturization for high-throughput automated analysis. Molecular applications of MagReSyn® Streptavidin microparticles include, protein immobilization and purification; nucleic acid capture assays and preparation of single-stranded DNA templates for e.g. SELEX and pyrosequencing. The immobilization of biotinylated antibodies may subsequently be used in e.g. immunoassay.