Learn More
ESI SOURCE SOLUTIONS MagReSyn Streptavidin MS, 2 ml, magnetic microparticles, > 25,000 pmoles/ml binding capacity (DNA), > 2.5 mg/ml binding capacity (Protein)
Supplier:  ESI SOURCE SOLUTIONS MRSTP002
Encompass
MagReSyn Streptavidin MS, 2 ml, magnetic microparticles, > 25,000 pmoles/ml binding capacity (DNA), > 2.5 mg/ml binding capacity (Protein)
Catalog No. NC2391689