Learn More
ESI SOURCE SOLUTIONS MagReSyn Streptavidin MS, 5 mL, Magnetic polymeric microparticle support, Binding capacity ≥2,500 pmoles/mg (biotinylated oligonucleotide)
Catalog No. NC2357174
Encompass
MagReSyn Streptavidin MS is a magnetic polymeric microparticle support designed for efficient isolation and immobilization of biotinylated biomolecules in proteomics workflows. This product allows for high loading capacity and minimizes non-specific interactions during mass spectrometry analysis. Its unique modifications chemically treat lysine and arginine residues on streptavidin, reducing contamination during enzyme digestion. It is ideal for applications like BioID that focus on on-bead digestion for mass spectrometry.
  • High streptavidin binding capacity for biomolecule penetration.
  • Reduces contamination from streptavidin peptides.
  • Facilitates miniaturization of experimental protocols.
  • Compatible with LysC and trypsin for enzymatic digestion.
  • Ensures high purity of target proteins.
  • Stable in pH range 3-10 and temperature range 4-60 °C.
  • Storage at 4-8 °C; do not freeze.
Catalog No. NC2357174 Supplier ESI SOURCE SOLUTIONS Supplier No. MRSTP005
MagReSyn Streptavidin MS is a magnetic polymeric microparticle support designed for efficient isolation and immobilization of biotinylated biomolecules in proteomics workflows. This product allows for high loading capacity and minimizes non-specific interactions during mass spectrometry analysis. Its unique modifications chemically treat lysine and arginine residues on streptavidin, reducing contamination during enzyme digestion. It is ideal for applications like BioID that focus on on-bead digestion for mass spectrometry.
  • High streptavidin binding capacity for biomolecule penetration.
  • Reduces contamination from streptavidin peptides.
  • Facilitates miniaturization of experimental protocols.
  • Compatible with LysC and trypsin for enzymatic digestion.
  • Ensures high purity of target proteins.
  • Stable in pH range 3-10 and temperature range 4-60 °C.
  • Storage at 4-8 °C; do not freeze.