Learn More
ESI SOURCE SOLUTIONS MagReSyn Zr-IMAC HP, Magnetic Microparticles, 10 mL volume, Zr4+ metal ion, 5-10 μm particle size, pH stability 2.5-12, temp stability 4-60 °C
Supplier:  ESI SOURCE SOLUTIONS MRZHP010.
Encompass
MagReSyn Zr-IMAC HP, Magnetic Microparticles, 10 mL volume, Zr4+ metal ion, 5-10 μm particle size, pH stability 2.5-12, temp stability 4-60 °C
Catalog No. NC3664548