Learn More
Medchemexpress LLC Magrolimab 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP990295MG
Encompass_Preferred
Magrolimab (Hu5F9-G4) is a humanized anti-CD47 antibody Magrolimab can block the don t eat me signal of CD47 thereby promoting macrophage mediated phagocytosis Magrolimab has antitumor activity in malignant bone tumors and breast cancer[1][2][3] /br
Catalog No. 50-003-25554