Learn More
Amsbio LLC MagSi-DNA allround COOH
Catalog No. 502041428
Encompass
MagSi-DNA allround beads are ideal for developing your own nucleic acid isolation and extraction methods, such as: - Isolation of genomic, mitochondrial, or viral DNA from whole blood, cell lysates, human, animal, or plant tissue; isolation of RNA - Isolation of genomic, plasmid, or phage DNA from bacterial cultures and bacteria from clinical samples (blood, stool, swabs, etc.) - Clean-up of DNA from enzymatic reactions (restriction digestions, ligations) or chromatin immunoprecipitation (ChIP) procedures to remove excess primers, nucleotides, enzymes, salts, buffers and other substances that are unwanted in downstream applications. | MagSi-DNA allround beads are magnetic silica beads with a nano-porous surface optimal for nucleic acid binding. Due to the superparamagnetic properties and size (1.2 um), the beads sediment slowly and typically collect within 2-3 minutes in a magnetic field. This makes them applicable for both manual and automated/robotic DNA isolation.
Catalog No. 50-204-1428 Supplier Amsbio LLC Supplier No. MD02020
MagSi-DNA allround beads are ideal for developing your own nucleic acid isolation and extraction methods, such as: - Isolation of genomic, mitochondrial, or viral DNA from whole blood, cell lysates, human, animal, or plant tissue; isolation of RNA - Isolation of genomic, plasmid, or phage DNA from bacterial cultures and bacteria from clinical samples (blood, stool, swabs, etc.) - Clean-up of DNA from enzymatic reactions (restriction digestions, ligations) or chromatin immunoprecipitation (ChIP) procedures to remove excess primers, nucleotides, enzymes, salts, buffers and other substances that are unwanted in downstream applications. | MagSi-DNA allround beads are magnetic silica beads with a nano-porous surface optimal for nucleic acid binding. Due to the superparamagnetic properties and size (1.2 um), the beads sediment slowly and typically collect within 2-3 minutes in a magnetic field. This makes them applicable for both manual and automated/robotic DNA isolation.