Learn More
Amsbio LLC MagSi-DNA mf
Catalog No. 502041441
Encompass
MagSi-DNA mf is ideal for developing your own application, such as: - Isolation of genomic, mitochondrial, or viral DNA from whole blood, cell lysates, human, animal, or plant tissue; isolation of RNA - Isolation of genomic, plasmid, or phage DNA from bacterial cultures and bacteria from clinical samples (blood, stool, swabs, etc.) - Clean-up of DNA from enzymatic reactions or chromatin immunoprecipitation (ChIP) | MagSi-DNA mf consists of magnetic silica beads with a highly dense core of magnetite. Due to their ferrimagnetic properties, the beads typically collect within 10 seconds in a magnetic field, but quickly loose magnetic remanence and are easily resuspended. The beads have minimal adhesion effects to plastics, making them perfectly suitable for use in microfluidic systems. The small particle size offers a very large active surface area for binding of nucleic acids, and the storage buffer has been optimized for maximum suspension time and ease of resuspension.
Catalog No. 50-204-1441 Supplier Amsbio LLC Supplier No. MD0200010010
MagSi-DNA mf is ideal for developing your own application, such as: - Isolation of genomic, mitochondrial, or viral DNA from whole blood, cell lysates, human, animal, or plant tissue; isolation of RNA - Isolation of genomic, plasmid, or phage DNA from bacterial cultures and bacteria from clinical samples (blood, stool, swabs, etc.) - Clean-up of DNA from enzymatic reactions or chromatin immunoprecipitation (ChIP) | MagSi-DNA mf consists of magnetic silica beads with a highly dense core of magnetite. Due to their ferrimagnetic properties, the beads typically collect within 10 seconds in a magnetic field, but quickly loose magnetic remanence and are easily resuspended. The beads have minimal adhesion effects to plastics, making them perfectly suitable for use in microfluidic systems. The small particle size offers a very large active surface area for binding of nucleic acids, and the storage buffer has been optimized for maximum suspension time and ease of resuspension.