Learn More
Antibodies Online MagSi-S Epoxy 3.0 Beads, 2ml
SDP
Catalog No. NC2751511
Encompass
PurposeSuperparamagnetic silica particles with epoxy modified surface. Intended for epoxy coupling chemistry to enzymes and other NH2 containing molecules.Bead Size - 3 µmCharacteristicsCoating chemistry: EpoxyBead concentration: 1 - 3 x 10^9 beads/mLComponentsMagnetic silica beads with activated surface
Catalog No. NC2751511 Supplier Antibodies Online Supplier No. ABIN1721106
PurposeSuperparamagnetic silica particles with epoxy modified surface. Intended for epoxy coupling chemistry to enzymes and other NH2 containing molecules.Bead Size - 3 µmCharacteristicsCoating chemistry: EpoxyBead concentration: 1 - 3 x 10^9 beads/mLComponentsMagnetic silica beads with activated surface