Learn More
Amsbio LLC MagSi-WCX
Supplier:  Amsbio LLC MD01023
Encompass
MagSi-WCX beads are magnetic silica beads coated with a weak cation exchange surface (WCX). The beads are intended for: - sample preparation and pre-fractionation prior to mass spectrometry (e.g. MALDI-TOF analysis) and HPLC - protein and peptide separation for multiple downstream applications, e.g. enzymatic assays - detergent removal - Fractionation of clinical samples e.g. serum, plasma, tissues, CSF, urine and cell lysate
Catalog No. 50-204-1575