Learn More
Magbio Genomics Inc MAGSTRIP STAND 1.5MLX12
SDP
Catalog No. NC2003790
Encompass
MagStrip Magnet Stand is designed to simplify the manual processing of magnetic bead separations for nucleic acid purification and clean up. The stand can accommodate either 12 standard 1.5 ml microcentrifuge tubes. The use of magnetic separation devices is essential in any paramagnetic bead-based DNA and RNA purification process. MAGBIO GENOMICS INC mfg# MBMS-12
Catalog No. NC2003790 Supplier Magbio Genomics Inc Supplier No. MBMS12
MagStrip Magnet Stand is designed to simplify the manual processing of magnetic bead separations for nucleic acid purification and clean up. The stand can accommodate either 12 standard 1.5 ml microcentrifuge tubes. The use of magnetic separation devices is essential in any paramagnetic bead-based DNA and RNA purification process. MAGBIO GENOMICS INC mfg# MBMS-12