Learn More
Edge Biosystems Inc MagDTR 96-Well Magnetic Separator
SDP
Catalog No. NC0199574
Encompass
MagDTR Magnetic Separator 96 features a unique recessed well design with bar magnets imbedded in an aluminum block that provides significant performance advantages in magnetic separation for 96-well applications.
Catalog No. NC0199574 Supplier Edge Biosystems Inc Supplier No. 57624
MagDTR Magnetic Separator 96 features a unique recessed well design with bar magnets imbedded in an aluminum block that provides significant performance advantages in magnetic separation for 96-well applications.