Learn More
Cayman Chemical MAHMA NONOate, CAS: 146724-86-9, 100 mg
SDP
Catalog No. 502896744
Encompass_Preferred
MAHMA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 1 minute and 3 minutes at 37°C and 22-25°C, respectively, (pH 7.4) to liberate 2 moles of NO per mole of parent compound.{1190,5437}
Catalog No. 50-289-6744 Supplier Cayman Chemical Supplier No. 82130100 MG
MAHMA NONOate is a NO donor. It spontaneously dissociates in a pH-dependent, first-order process with a half-life of 1 minute and 3 minutes at 37°C and 22-25°C, respectively, (pH 7.4) to liberate 2 moles of NO per mole of parent compound.{1190,5437}