Learn More
Enzo Life Sciences MAHMA NONOate (5mg). CAS: 146724-86-9
SDP
Supplier:  Enzo Life Sciences ALX4300155005
Encompass_Preferred
Nitric oxide (NO) donor. The half-life time of MAHMA NONOate is 2.7 min. at 22°C in 0.1M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0. Alternative name: NOC-9, MAHMA/NO, (Z)-1-(N-Methyl-N-[6-(N-methylammoniohexyl)amino])-diazen-1-ium-1,2-diolate. Purity: ≥98%. Appearance: White to off-white crystalline solid. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9141