Learn More
MSC Mahr 1004 Compramess Dial Comparator Gage, 0.0001 in. Graduation, 0.005 in. Range, 1 ea.
Catalog No. 5000517006
Encompass
This dial comparator gage provides high-precision dimensional measurements, featuring a 0.0001 in. graduation and a 0.005 in. maximum measurement range. Engineered for durability and sensitivity, it incorporates a jeweled movement and a high-precision ball guide to ensure minimal hysteresis and reliable performance.
  • Shockproof movement for long-term reliability
  • Large, high-contrast white dial face for clear readability
  • Adjustable tolerance markers for efficient measurement monitoring
  • Measuring spindle and mounting shank constructed from hardened stainless steel
  • Spindle can be retracted using a screw-in cable release or a lifting knob
  • Resistant to lateral forces acting on the measuring spindle
  • Includes adapter bush (8 mm to 3/8 in.) and a protective case
Catalog No. 50-005-17006 Supplier MSC Supplier No. 06467542
This dial comparator gage provides high-precision dimensional measurements, featuring a 0.0001 in. graduation and a 0.005 in. maximum measurement range. Engineered for durability and sensitivity, it incorporates a jeweled movement and a high-precision ball guide to ensure minimal hysteresis and reliable performance.
  • Shockproof movement for long-term reliability
  • Large, high-contrast white dial face for clear readability
  • Adjustable tolerance markers for efficient measurement monitoring
  • Measuring spindle and mounting shank constructed from hardened stainless steel
  • Spindle can be retracted using a screw-in cable release or a lifting knob
  • Resistant to lateral forces acting on the measuring spindle
  • Includes adapter bush (8 mm to 3/8 in.) and a protective case