Learn More
MSC Mahr Digital Electronic Drop Indicator, 25 mm Max, 0.001 mm Resolution, Flat Back
Catalog No. 5000529296
Encompass
This digital electronic drop indicator offers precise measurement with a maximum range of 25 mm (1 inch). Featuring a flat back design and a high contrast LCD, it provides clear readings and comes calibrated for immediate use in various applications.
  • High contrast LCD
  • Includes battery
  • Includes instruction manual
  • Calibrated for precision measurement
Catalog No. 50-005-29296 Supplier MSC Supplier No. 18870816
This digital electronic drop indicator offers precise measurement with a maximum range of 25 mm (1 inch). Featuring a flat back design and a high contrast LCD, it provides clear readings and comes calibrated for immediate use in various applications.
  • High contrast LCD
  • Includes battery
  • Includes instruction manual
  • Calibrated for precision measurement