Learn More
MSC Mahr Digital Indicator, 12.5 mm / 0.5 in Range, 0.01 mm Resolution, IP52
Catalog No. 5000514620
Encompass
The digital indicator provides precise length measurements for various industrial settings, including production, quality control, and inspection areas. It features a high-contrast LCD display, robust construction with hardened stainless steel components, and offers selectable resolutions. With IP52 protection, it ensures reliable operation and supports multiple data output options for seamless integration.
  • High-contrast LCD display for clear readability.
  • Mounting shank and measuring spindle are made of hardened stainless steel.
  • Robust construction for enhanced durability.
  • IP52 protection against dust and splashing water.
  • Data output options: Digimatic, Opto RS-232C, or USB.
  • Automatic ON/OFF function conserves battery life.
  • Ability to change the measuring direction.
  • Lock function secures individual buttons during operation.
Catalog No. 50-005-14620 Supplier MSC Supplier No. 70771597
The digital indicator provides precise length measurements for various industrial settings, including production, quality control, and inspection areas. It features a high-contrast LCD display, robust construction with hardened stainless steel components, and offers selectable resolutions. With IP52 protection, it ensures reliable operation and supports multiple data output options for seamless integration.
  • High-contrast LCD display for clear readability.
  • Mounting shank and measuring spindle are made of hardened stainless steel.
  • Robust construction for enhanced durability.
  • IP52 protection against dust and splashing water.
  • Data output options: Digimatic, Opto RS-232C, or USB.
  • Automatic ON/OFF function conserves battery life.
  • Ability to change the measuring direction.
  • Lock function secures individual buttons during operation.