Learn More
MSC Mahr Horizontal Dial Test Indicator, 0.0150 in Max, 0-15-0 Dial, 0.001 in Accuracy
Catalog No. 5000536241
Encompass
This horizontal dial test indicator features a maximum measurement of 0.0150 in and a 0-15-0 dial reading. It offers an accuracy of 0.001 in. The dial has a diameter of 1-1/8 in, with a green face and a graduation of 0.001 in. It includes a ball diameter of 0.0787 in and a contact point length of 0.5700 in (14.50 mm).
  • Ball bearing mounted
  • Mineral glass viewing panel
  • Matt chrome-plated finish
  • Sealed design
  • Automatic adjustment
  • Friction clutch overload protection
  • High-contrast dial face
  • Anti-magnetic model
  • Shockproof measuring mechanism
  • Includes 2 mm stylus, 800a3/8 mounting shaft, case, instruction manual, and spanner
Catalog No. 50-005-36241 Supplier MSC Supplier No. 09685108
This horizontal dial test indicator features a maximum measurement of 0.0150 in and a 0-15-0 dial reading. It offers an accuracy of 0.001 in. The dial has a diameter of 1-1/8 in, with a green face and a graduation of 0.001 in. It includes a ball diameter of 0.0787 in and a contact point length of 0.5700 in (14.50 mm).
  • Ball bearing mounted
  • Mineral glass viewing panel
  • Matt chrome-plated finish
  • Sealed design
  • Automatic adjustment
  • Friction clutch overload protection
  • High-contrast dial face
  • Anti-magnetic model
  • Shockproof measuring mechanism
  • Includes 2 mm stylus, 800a3/8 mounting shaft, case, instruction manual, and spanner