Learn More
MSC Mahr Parallel Chisel Probe, 10 μm Radius, 90° Sapphire Chisel, for Pocket Surf IV
Catalog No. 5000530642
Encompass
This parallel chisel probe is designed for gaging sharp edges or small outer diameters where the probe is positioned perpendicularly (in 90° or 270° position) to the axis of traverse. It can also be used with the EAS-2421 Vee fixture for outer diameters smaller than 6.35 mm.
  • Suitable for measuring sharp edges.
  • Ideal for measuring small outer diameters.
  • Can be used in a perpendicular orientation to the traverse axis (90° or 270° position).
  • Compatible with EAS-2421 Vee fixture for very small outer diameters.
Catalog No. 50-005-30642 Supplier MSC Supplier No. 66010125
This parallel chisel probe is designed for gaging sharp edges or small outer diameters where the probe is positioned perpendicularly (in 90° or 270° position) to the axis of traverse. It can also be used with the EAS-2421 Vee fixture for outer diameters smaller than 6.35 mm.
  • Suitable for measuring sharp edges.
  • Ideal for measuring small outer diameters.
  • Can be used in a perpendicular orientation to the traverse axis (90° or 270° position).
  • Compatible with EAS-2421 Vee fixture for very small outer diameters.