Learn More
Cayman Chemical mAIhydrochlorIde 10mg
SDP
Catalog No. 502954239
Encompass_Preferred
A psychoactive aminoindan which can be found in recreational drug mixtures; potently inhibits serotonin uptake (IC50 = 212 nM) and stimulates its release without inducing neurotoxicity
Catalog No. 50-295-4239 Supplier Cayman Chemical Supplier No. 1389510 MG
A psychoactive aminoindan which can be found in recreational drug mixtures; potently inhibits serotonin uptake (IC50 = 212 nM) and stimulates its release without inducing neurotoxicity